Print…
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 800)
Fornecedor: AAT Bioquest
AATB100520N1EA 1390 EUR
AATB100520N1 AATB100520N0
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 800)
Anticorpos
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 800
Epitope:
Host: Mouse
Isotype:
Reactivity: Human