Print…
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 633)
Fornecedor: AAT BIOQUEST
116100E0 116100E1
AATB116100E0EA 371 EUR
AATB116100E0 AATB116100E1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 633)
Anticorpos
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 633
Epitope:
Host: Mouse
Isotype:
Reactivity: Human