Print…

Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP (Peridinin-Chlorophyll Protein Complex))

Fornecedor: AAT BIOQUEST
101921T0 101921T1 101921T2
AATB101921T0EA 271 EUR
AATB101921T0 AATB101921T1 AATB101921T2
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (PerCP (Peridinin-Chlorophyll Protein Complex))
Anticorpos
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR