Print…

Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 594)

Fornecedor: AAT BIOQUEST
100210C0 100210C1
AATB100210C0EA 371 EUR
AATB100210C0 AATB100210C1
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 594)
Anticorpos
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR