Print…
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 488)
Fornecedor: AAT BIOQUEST
10032050 10032051
AATB10032050EA 371 EUR
AATB10032050 AATB10032051
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (iFluor® 488)
Anticorpos
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kDa member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependent)s.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: iFluor® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: Human