Print…
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 488)
Fornecedor: AAT BIOQUEST
10021050 10021051
AATB10021050EA 371 EUR
AATB10021050 AATB10021051
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10] (iFluor® 488)
Anticorpos
RPA-2.10 is an anti-human monoclonal antibody that is specific for the CD2 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: iFluor® 488
Epitope:
Host: Mouse
Isotype:
Reactivity: Human