Print…
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 430)
Fornecedor: AAT BIOQUEST
10161030 10161031
AATB10161030EA 371 EUR
AATB10161030 AATB10161031
Anti-CD16a Mouse Monoclonal Antibody [clone: 3G8] (iFluor® 430)
Anticorpos
The 3G8 monoclonal antibody recognises human CD16a, a 50 to 65 kDa transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: iFluor® 430
Epitope:
Host: Mouse
Isotype:
Reactivity: Human