Print…

Anti-Mouse IgG Goat Polyclonal Antibody (CF®583R)

Fornecedor: Biotium
20903-500UL 20903-50UL
20903-500ULEA 321 EUR
20903-500UL 20903-50UL
Anti-Mouse IgG Goat Polyclonal Antibody (CF®583R)
Anticorpos
This is a highly cross-adsorbed goat anti-mouse IgG (H L) secondary antibody labeled with our bright and photostable CF® Dyes and other labels. To minimise cross-reactivity, the antibody has been adsorbed against bovine, chicken, goat, guinea pig, Syrian hamster, horse, human, rabbit, rat and sheep serum proteins.

  • Highly cross-adsorbed for specific staining with minimal background
  • Suitable for western, immunofluorescence, and immunohistology
  • Conjugation: CF®583R

Type: Secondary
Antigen: Mouse IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR