Print…
Analog Thermostatised Baths
Fornecedor: SUMINISTROS GRUPO ESPER S L
10000-01107 10000-01106 10000-01105 10000-01101
OVAN10000-01107EA 1280 EUR
OVAN10000-01107 OVAN10000-01106 OVAN10000-01105 OVAN10000-01101
Analog Thermostatised Baths
Banhos Banhos-maria sem agitação
Thermostatised baths with inner tank made of stainless steel AISI 304 and metallic housing protected with epoxy paint of great resistance to aggressive surroundings.
  • Temperature controlled by means of an hydraulic thermostat, for water and oil
  • Water drainage faucet and handles for a easy transport
Heating resistance embedded made of special alloy of stainless steel, heat and corrosion resistant. Temperature thermostat
synchronised with the 'on' indicator lamp. Safety cut out thermostat with indicator lamp. The unit will cut out if there is insufficient liquid or if the temperature exceeds the maximum admissible temperature.
Certificações: IP 52 protection class.
Fornecimento: Includes stainless steel cover-resistance.