Print…
HYGEN™ Microfibre Wet Mop Pads
Fornecedor: Rubbermaid Commercial Products
R034556 R050650 R050647
141-0144EA 24.7 EUR
141-0144 129-0949 129-0948
HYGEN™ Microfibre Wet Mop Pads
Esfregonas
HYGEN™ microfibre wet mop pads are constructed of premium 16-split nylon/polyester blend microfibre that provides high-efficacy wet mopping preformance.
  • Patented zig-zag scrubbing strips for cleaning tough, dried-on stains
  • Velcro attachment system
  • Advanced hook-and-loop pad holds up under harsh laundering conditions and attaches pads more securely
  • Durable up to 500 launderings* (up to 200 with bleach)
  • Bleach-safe in hot and cold water: Certified to tolerate CDC hot and cold water laundering requirements**
The microfibre wet pad is designed for general-purpose use, and features colour-coded tabs to prevent cross-contamination. The microfibre scrubber wet pad features vertical polypropylene scrubber stripes that help remove stubborn spots and clean into tile grout lines. The microfibre high-absorbency wet pad addresses big spills by absorbing up to 710 ml of liquid.
Tested pathogen removal with HYGEN™ microfibre wet mop pads.
Removes 99,9% or more of tested viruses and bacteria, with water only.***
Tested viruses: Feline calicivirus (surrgate for norovirus), common human coronavirus OC43.
Tested bacteria: Methicillin-Resistant Staphylococcus Aureus (MRSA), Clostridioides difficile (C. diff), pseudomonas aeruginosa.
Not tested on COVID-19.
*Based on internal testing without bleach.
**Always use cleaning chemicals or disinfectants as directed by a facility manager.
***Based on third-party testing on VCT surface with water only. EPA est. No 92100-CHM-1. Claim does not apply to R034556.