Print…
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 350)
Fornecedor: AAT BIOQUEST
10280010 10280011
AATB10280010EA 371 EUR
AATB10280010 AATB10280011
Anti-CD28 Mouse Monoclonal Antibody [clone: CD28.2] (iFluor® 350)
Anticorpos
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 350
Epitope:
Host: Mouse
Isotype:
Reactivity: Human