Print…

Ethyl phenylpropiolate, Sigma-Aldrich®

Fornecedor: Merck

Sinônimos: Phenylpropiolic acid ethyl ester, Ethyl 3-phenylprop-2-ynoate, Ethyl phenylacetylenecarboxylate

Nível de qualidade M-Clarity™ = MQ100

E45309-25G E45309-5G
SIALE45309-25GEA 203 EUR
SIALE45309-25G SIALE45309-5G
Ethyl phenylpropiolate, Sigma-Aldrich®
Ethyl phenylpropiolate
Fórmula: C₁₁H₁₀O₂
Peso Molecular: 174.2 g/mol
Ponto de ebulição: 260
Ponto de Fusão: 60
Densidade: 1.05 g/cm³ (20 °C)
Número MDL : MFCD00009185
Número CAS: 2216-94-6
EINECS: 218-703-8

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR