L(+)-Glutamato de sódio monidratado, Sigma-Aldrich®
Fornecedor: Merck
Sinônimos:
(S)-(+)-2-Aminopentanedioic acid sodium salt monohydrate, Sodium L-Glutamate Monohydrate, L(+)-Glutamic acid sodium salt monohydrate, L(+)-2-Aminopentanedioic acid sodium salt monohydrate, (S)-(+)-Glutamic acid sodium salt monohydrate, L(+)-Glutamic acid monosodium salt monohydrate, (S)-(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-Sodium glutamate monohydrate, L(+)-α-Glutamic acid sodium salt monohydrate, L(+)-1-Aminopropane-1,3-dicarboxylic acid sodium salt monohydrate, (S)-(+)-α-Glutamic acid sodium salt monohydrate
Nível de qualidade M-Clarity™ = MQ200
Fórmula:
C₅H₈NNaO₄·H₂O Peso Molecular: 187.13 g/mol Ponto de Fusão: 225…240 °C Ponto de Inflamação: >225 °C Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00150138 Número CAS: 6106-04-3 EINECS: 240-313-1 Merck Index: 13,06278 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...