Print…

Indo-1 AM ≥95%, Grau ultra puro

Fornecedor: AAT BIOQUEST
Atenção

21036 21032
AATB21036EA 273 EUR
AATB21036 AATB21032
Indo-1 AM ≥95%, Grau ultra puro
Indo-1 AM

Calcium measurements are critical for numerous biological investigations.

Fórmula: C₄₇H₅₁N₃O₂₂
Número MDL : MFCD00036955
Número CAS: 112926-02-0
ADR: 5,II

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR