Print…
ReadiUse™ Disposable PD-10 Desalting Column
Fornecedor: AAT BIOQUEST
60504 60505
AATB60504EA 210 EUR
AATB60504 AATB60505
ReadiUse™ Disposable PD-10 Desalting Column
Cromatografia, Colunas
PD-10 Desalting columns contain sephadex G-25 resin for rapid buffer exchange, desalting, and removal of small contaminants (salts, dyes, radioactive labels) from samples using gravity flow or centrifugation.
  • Efficient contaminant removal
  • Excellent sample recovery
  • Dialysis alternative
Atenção: For research use only.