Screen Quest™ Live Cell cAMP Assay Service Pack
Fornecedor: AAT BIOQUEST
36382
36383
36384
AATB36382EA
2080
EUR
AATB36382
AATB36383
AATB36384
Screen Quest™ Live Cell cAMP Assay Service Pack
Ensaios
Ensaios celulares
G protein coupled receptors (GPCR) are one of the largest receptor classes targeted by drug discovery programs.
- Extremely sensitive detection
- Contains all necessary components
- No lysis step required
- Robust and adaptable protocol
- Longer fluorescence lifetime
Atenção: For research use only.