Rhod-2 AM (Bromide) ≥95%, Grau ultra puro
Fornecedor: AAT BIOQUEST
|
Atenção
|
Sinônimos:
Rhod-2 AM ester, 1-[2-Amino-5-(3-dimethylamino-6-dimethylammonio-9-xanthenyl)phenoxy]-2-(2-amino-5-methylphenoxy)]ethane-N,N,N',N'-tetraacetic acid, tetraacetoxy methyl ester . Br
21062
21063
21064
AATB21062EA
307
EUR
AATB21062
AATB21063
AATB21064
Rhod-2 AM (Bromide) ≥95%, Grau ultra puro
Rhod-2 AM (Bromide)
Calcium measurement is critical for numerous biological investigations.
Fórmula:
C₅₂H₅₉BrN₄O₁₉ Peso Molecular: 1124.0 g/mol |
Número CAS:
145037-81-6 ADR: 5 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...