Print…

Fura 2-AM ≥95%, Grau ultra puro

Fornecedor: AAT BIOQUEST
Atenção

Sinônimos: FURA-2 pentakis(acetoxymethyl) ester, 1-[2-(5-Carboxyoxazol-2-yl)-6-aminobenzofuran-5-oxy]-2-(2'-amino-5'-methyl-phenoxy)ethane-N,N,N',N'-tetraacetic acid, pentaacetoxymethyl ester, FURA-2 acetoxymethyl ester, Fura-2 AM Ester

21021 21023
AATB21021EA 230 EUR
AATB21021 AATB21023
Fura 2-AM ≥95%, Grau ultra puro
Fura 2-AM

Among the ratiometric calcium indicators, Fura-2 and Indo-1 are most commonly used.

Fórmula: C₄₄H₄₇N₃O₂₄
Peso Molecular: 1001.857 g/mol
Número MDL : MFCD00036976
Número CAS: 108964-32-5
ADR: 5 II

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR