bisBenzimide H 33342 trihydrochloride ≥95%, Grau ultra puro
Fornecedor: AAT BIOQUEST
Sinônimos:
Hoechst 33342, bisBenzimide, 2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342
17530
17533
AATB17530EA
152
EUR
AATB17530
AATB17533
bisBenzimide H 33342 trihydrochloride ≥95%, Grau ultra puro
bisBenzimide H 33342 trihydrochloride
The Hoechst stains are a family of fluorescent stains for labeling DNA in fluorescence microscopy.
- Bright Fluorescence
- 95% Purity
- Useful For Labeling Nuclei/Mitochondria
- DAPI Alternative
Fórmula:
C₂₇H₂₈N₆O·3HCl·xH₂O Peso Molecular: 561.93 g/mol (anhy) |
Número MDL :
MFCD00012678 Número CAS: 875756-97-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...