Print…
Thermal Grip Gloves - Latex
Fornecedor: Portwest
A140BKRL A140BKRM A140BKRS A140BKRXL A140BKRXS A140BKRXXL
PRTWA140BKRLEA 38 EUR
PRTWA140BKRL PRTWA140BKRM PRTWA140BKRS PRTWA140BKRXL PRTWA140BKRXS PRTWA140BKRXXL
Thermal Grip Gloves - Latex
Luvas
The thermal grip gloves are perfect for construction, transportation, refuse collection, maintenance, and local authority work.
  • Specially designed for use in cold conditions
  • Palm dipped to increase dexterity and ventilation
  • Crinkle latex grip offers excellent grip
  • Warm 10 gauge acrylic liner for cold protection
  • Breathable seamless liner
  • Ergonomically designed for comfort and ease of use
  • ANSI cut level A1
This glove has a warm acrylic 10 gauge liner and is ideal for more heavy-duty, outdoor tasks or those people working in cold conditions. Retail tag aids presentation for retail sales.
Colour: Black
Certificações: EN ISO 21420: 2020 Dexterity 4, EN 388: 2016 + A1: 2018 1141X, EN 511: 2006 X1X. ANSI/ISEA 105: 2016 CUT Level A1. CE certified.