Print…
Foam Nitrile Bamboo Gloves, NPR15
Fornecedor: Portwest
AP10K8RL AP10K8RM AP10K8RS AP10K8RXL AP10K8RXS AP10K8RXXL
112-6600EA 71.6 EUR
112-6600 112-6601 112-6602 112-6603 112-6604 112-6605
Foam Nitrile Bamboo Gloves, NPR15
Luvas
Extremely comfortable with excellent breathability, these gloves feature a 15-gauge liner constructed from bamboo viscose and nylon.
  • Made using a sustainable bamboo liner
  • Nitrile foam coating for excellent grip in wet and dry conditions
  • Excellent durability and grip in wet and dry conditions
  • 100% latex-free
  • 15 gauge liner for extra dexterity
  • Palm dipped to increase dexterity and ventilation
Bamboo is naturally anti-bacterial, hypoallergenic, and has resistance to UV light. The soft nitrile foam coated palm provides exceptional grip in wet and dry conditions. Perfect for a wide range of industries.
Colour: Black
Material: Bamboo/nylon/polyester
Certificações: EN ISO 21420: 2020 Dexterity 5, EN 388: 2016 + A1: 2018 4121X. CE certified.