Print…
VWR® Euro Containers, 100% Recycled Polypropylene
Fornecedor: VWR Collection
216-2997EA 10.7 EUR
216-2997 216-2998 216-2999 216-3000 216-3001
VWR® Euro Containers, 100% Recycled Polypropylene
Caixas Caixas de arrumação
This range of euro containers are made entirely from recycled polypropylene and adhere to the guidelines of the 'Der Blaue Engel' ecological label. Suitable for use in industry, handling, and automated transportation, they remain resilient to impacts and temperature variations. They can be stacked and nested without the risk of falling.
  • Solid stackable and palletisable storage bin made of polypropylene
  • Compliant with european standard for easy storage and transportation
  • Easy handling with open handles
  • Manufactured from recycled polypropylene
  • Lightweight and durable
  • Heights from 120 to 320 mm
This range, of professional quality and more environmentally friendly, is designed to meet intensive and daily usage needs. Adhering to the european standard dimensions ensures streamlined exchanges and logistics.
This series of euro container is entirely manufactured from recycled black polypropylene. Each model is equipped with two sturdy ergonomic handles that facilitate handling, making it easy to grasp and move the european bins. Despite being made from recycled polypropylene, they remain resilient to impacts and temperature fluctuations. They can be safely stacked and nested without the risk of toppling.