Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™
Fornecedor: THERMO FISHER DIAGNOSTICS
Quanti-Cult Plus™ Pseudomonas aeruginosa ATCC™ 9027™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial ready for rehydration and use in quality control procedures.
Embalagem: Includes 10 preserved organism vials and 10 rehydration vials.