Chitosan, pó
Fornecedor: Spectrum Chemical
Chitosan, Powder is a polysaccharide made by treating shrimp and other crustacean shells with the alkali sodium hydroxide. It can be used in agriculture as a seed treatment and to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Chitosan can be used in agriculture as a seed treatment and biopesticide to fight off fungal infections and is often used in the wine making industry as a chemical to prevent spoilage. In medicine it can be used in bandages to reduce bleeding and as an antibacterial agent.
Temperatura de Armazenamento:
Ambiente |
Número MDL :
MFCD00161512 Número CAS: 9012-76-4 |
Especificações dos Resultados do Teste
Synonyms | β-(1,4)-2-Amino-2-deoxy-D-glucose; Deacetylated Chitin; Poly(D-glucosamine) |
Form | Powder |
Deacetylation | Min. 90.0 % |
Moisture | Max. 10.0 % |
Ash | Max. 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...