Print…
Magne Protein A Beads, 20% Slurry
Fornecedor: Promega
G8781 G8782 G8783
PROMG8781EA 149 EUR
PROMG8781 PROMG8782 PROMG8783
Magne Protein A Beads, 20% Slurry
Esferas Esferas magnéticas
Magne Protein G and Magne Protein A Beads are magnetic affinity beads with high specificity and capacity for purification of immunoglobulins from cell culture media, ascites and serum. Iron encapsulated in macroporous cellulose with low nonspecific binding.
  • High specificity, high capacity magnetic affinity beads for antibody capture
  • Binding capacities of >25mg/ml depending on antibody species, isotype
  • Minimise purification losses and increase sample throughput with exceptional magnetic properties
Magne Protein G and Magne Protein A Beads are magnetic affinity beads with high specificity and high capacity for purification of immunoglobulins from cell culture media, ascites and serum samples. These paramagnetic beads are composed of iron encapsulated in macroporous cellulose with low nonspecific binding. The magnetic beads use a novel attachment chemistry to immobilise recombinant Protein G or Protein A protein molecules in the same orientation on the surface of the bead. The oriented attachment is known to improve the functionality of immobilised proteins. These beads offer a convenient method for achieving high purity and high recovery of monoclonal and polyclonal antibodies from a variety of biological samples. The superb magnetic properties of Magne Protein G and Magne Protein A Beads allow rapid and efficient capture of antibodies either with manually processed samples or in a high-throughput manner using the Promega ReliaPrep LV 32 HSM Instrument or a robotic platform such as the Beckman Coulter Biomek FX.