Print…
Cleanroom gloves, polyisoprene, sterile, Biogel® PI Tech Indicator® underglove
Fornecedor: MOLNLYCKE HEALTH CARE
4441655 4441660 4441665 4441670 4441675 4441680 4441685 4441690
112-6362EA 708 EUR
112-6362 112-6363 112-6372 112-6373 112-6374 112-6375 112-6376 112-6377
Cleanroom gloves, polyisoprene, sterile, Biogel® PI Tech Indicator® underglove
Luvas
These sterile, blue cleanroom undergloves with beaded cuffs and curved fingers are made to be worn in combination with the Biogel® PI Tech overglove to form a Breach Indication System. The gloves are suitable for use in aseptic and Class 100 (ISO 5)/EU GMP Grade A environments.
  • Blue underglove can be worn together with Biogel® PI Tech to form a Breach Indication System
  • Exceed industry standard for force at break, tensile strength, and elongation
  • Tested and cleared for use with chemotherapy agents
  • Synthetic polyisoprene
  • Biogel® hydrogel polymer coating for easy donning
  • Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
Ideal for use in controlled environments, in medical and research laboratories, in the pharma industry, as well as in biotechnology.
Certificações: EN 455, ASTM D3577, ISO 10282, EN 374, EN 16523-1, PPE Cat. III, AQL 0,65, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, ASTM F1671, Allergenicity ISO 10993, Pyrogenicity ASTM D7102, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420.
Embalagem: One pair per pack, in a high quality polyethylene inner wrap, packed into a film pack. 25 pairs per inner LDPE polybag f; 2 inner LDPE polybags are packed in an outer LDPE polybag. Four outer polybags per transit case, total of 200 pairs.