Print…
Quanti-CultiControl freeze-dried microorganism strains derived from ATCC® cultures
Fornecedor: LIOFILCHEM
89501 89502 89503 89506 89507 89508 89513 89514 89516 89517 89518 89519 89521 89522 89523 89524 89525 89526 89527 89528 89530 89531 89533 89535 89534 89537 89538
LIOF89501EA 184 EUR
LIOF89501 LIOF89502 LIOF89503 LIOF89506 LIOF89507 LIOF89508 LIOF89513 LIOF89514 LIOF89516 LIOF89517 LIOF89518 LIOF89519 LIOF89521 LIOF89522 LIOF89523 LIOF89524 LIOF89525 LIOF89526 LIOF89527 LIOF89528 LIOF89530 LIOF89531 LIOF89533 LIOF89535 LIOF89534 LIOF89537 LIOF89538
Quanti-CultiControl freeze-dried microorganism strains derived from ATCC® cultures
Testes para microbiologia Testes de Microrganismos
Quanti-CultiControl are lyophilized, quantitative microorganism preparations to be used in industrial laboratories for Quality Control purposes. Processed as directed, these preparations provide a challenge of < 100 CFU per 0.1 mL, a concentration suitable for several pharmaceutical applications, including growth promotion testing of culture media to be employed in sterility testing
  • Derived from the American Type Culture Collection (ATCC®)
  • Suitable for the Growth Promotion Test recommended by the international Pharmacopoeiae
  • No need of intermediate dilution steps, since each rehydrated suspension generates less than 100 UFC for 0.1 ml inoculum
  • 24 different Recommended Growth Methods are available (see the technical document in the documentation sheet)
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark and the ATCC catalog marks are trademarks of ATCC. Liofilchem® s.r.l. is licensed to use these trademarks and to sell products derived from ATCC® cultures
Certificações: Suitable for the Growth Promotion Test recommended by the International Pharmacopoeiae.
Embalagem: 1 vial containing 1 pellet + 1 rehydration fluid vial. Each suspension (1 ml) allows 10 inocula.