Print…
Industrial natural ruber latex gloves, AlphaTec® 87-950 series
Fornecedor: Ansell
87-950/8 87-950/9 87-950/10
ANSE87-950/8EA 49 EUR
ANSE87-950/8 ANSE87-950/9 ANSE87-950/10
Industrial natural ruber latex gloves, AlphaTec® 87-950 series
Luvas
Extra heavy-duty natural rubber glove with flocked lining.
  • Extra heavy-duty black glove
  • Durable design, with 0,73 mm/29 mil thickness
  • 100% cotton flock lining
  • AQL 1.5
Engineered to be 50% thicker than standard flock lined gloves, these heavy-duty gloves offer extremely high resistance to many ketones, salts, detergents, alcohols, alkalies, and fats. They provide excellent resistance to mechanical hazards plus increased chemical protection. Chlorinated with 100% cotton flock lining to help absorb perspiration.