Print…
Masterflex® High-Efficiency Absolute-Rated Filter Bags, Polypropylene, Avantor®
Fornecedor: Avantor Fluid Handling
Masterflex®
MFLX01531-60EA 61 EUR
MFLX01531-60 MFLX01531-70 MFLX01531-74 MFLX01531-68
Masterflex® High-Efficiency Absolute-Rated Filter Bags, Polypropylene, Avantor®
Filtros Bag Filter Systems
Wide range of options for temperature, flow rate, and filtration quality.
  • Constructed of polypropylene (PP)
  • FDA-compliant
Designed for 98% removal efficiency at the rated pore size, these bags offer three to four times the dirt-holding capacity of standard bags. In addition, they are FDA-compliant with respect to all regulations governing food-grade materials. These polypropylene absolute-rated filter bags are rated for a temperature of 200 °F (93 °C) maximum.