Print…
Masterflex® Bag Filters for Polypropylene Bag Filter Housings, Avantor®
Fornecedor: Avantor Fluid Handling
Masterflex®
MFLX01516-70EA 18 EUR
MFLX01516-70 MFLX01519-00 MFLX01516-50 MFLX01516-94 MFLX01516-72 MFLX01516-30 MFLX01516-32 MFLX01516-10 MFLX01516-76 MFLX01516-12 MFLX01519-02 MFLX01519-06 MFLX01516-08 MFLX01516-80 MFLX01516-40 MFLX01516-84 MFLX01519-12 MFLX01516-62 MFLX01516-20 MFLX01516-66 MFLX01516-00 MFLX01516-44 MFLX01519-16 MFLX01516-22 MFLX01516-88 MFLX01516-24 MFLX01516-02 MFLX01516-04 MFLX01516-26 MFLX01516-28 MFLX01516-06
Masterflex® Bag Filters for Polypropylene Bag Filter Housings, Avantor®
Filtros Bag Filter Systems
Conveniently collect all filtered particles within the bag.
  • Choose from a wide range of bag materials suitable for your application
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.