Print…
Automation tips for Hamilton® Microlab® Star Line and Nimbus®, Axygen
Fornecedor: Corning
HT-50-CBK-HTRS HT-1000CBKHTRS HT-10-CBK-HTR HT-10-CBK-HTRS HT-300-CBK-HTR HT-300-CBKHTRS HT-50-CBK-HTR HTF1000CBKHTRS HTF-10-CBKHTRS HTF-300CBKHTRS HTF-50-CBKHTRS HT-10-CBK-STK HT-300-CBK-STK HT-50-CBK-STK HT10CBKHTR5 HT1000CHTRS5 HT10CBKHTRS5 HT50CBKHTR5 HT300CBKHTR5 HT1000CBKHTR5 HT1000CBKHTRS5 HT10CHTR5 HT10CHTRS5 HT50CHTR5 HT300CHTR5 HT300CHTRS5 HT1000CHTR5 HT50CBKHTRS5
731-0423EA 297.66 EUR
731-0423 731-0416 731-0417 731-0418 731-0420 613-3138 731-0422 731-0425 731-0426 731-0427 731-0428 731-0419 731-0421 731-0424 AXYGHT10CBKHTR5 AXYGHT1000CHTRS5 AXYGHT10CBKHTRS5 AXYGHT50CBKHTR5 AXYGHT300CBKHTR5 AXYGHT1000CBKHTR5 AXYGHT1000CBKHTRS5 AXYGHT10CHTR5 AXYGHT10CHTRS5 AXYGHT50CHTR5 AXYGHT300CHTR5 AXYGHT300CHTRS5 AXYGHT1000CHTR5 AXYGHT50CBKHTRS5
Automation tips for Hamilton® Microlab® Star Line and Nimbus®, Axygen
Pontas de pipetas
Axygen brand automation tips are compatible with Hamilton® Microlab® Star and Nimbus® robotic systems.
  • Conductive black or clear polypropylene
  • RNase-/DNase-/human gDNA-free
  • Nonpyrogenic
Tips are available in 4 sizes (10, 50, 300, and 1000 µl) and are packaged in either traditional hanging tip racks ('frame' style) or deck-seating nested tip racks. As with all Axygen robotic tips, they are manufactured to stringent specifications under strict process control and assembled via automation to ensure consistent performance and quality.
Informações de encomenda: Tips are also available in five tray blister packs with barcoded trays.