Print…
Overalls, Biztex™ Pro ST90, type 5/6
Fornecedor: Portwest
ST90WHRM ST90WHRL ST90WHRXL ST90WHRXXL ST90WHRXXXL
PRTWST90WHRMEA 421 EUR
PRTWST90WHRM PRTWST90WHRL PRTWST90WHRXL PRTWST90WHRXXL PRTWST90WHRXXXL
Overalls, Biztex™ Pro ST90, type 5/6
Uniformes
The spacious design guarantees a good fit and will not limit job performance.
  • Reusable and machine washable up to 5 times
  • Ultra-comfort and breathability
  • Excellent liquid and particulate barrier protection
  • Lightweight and comfortable
  • Elasticated hood, ankles, cuffs and waist
  • Sealable zip storm flap for extra protection
  • Ergonomically shaped hood for a perfect fit
  • Two-way zip for quick and easy access
Made from a comfortable and durable polyester tricot warp knit and laminated with a low permeability polyethylene film. Practical features include an elasticated hood, ankles, cuffs and waist, along with a two-way front zip and sealable front flap.
Shell fabric: BizTex Pro 75g
Certificações: EN ISO 13982 Type 5B. EN 13034 Type 6B. EN 14126; Cat. III