Print…
Klebsiella aerogenes (formerly Enterobacter aerogenes) WDCM 00175 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Fornecedor: Merck
VT001753-10EA VT001754-10EA VT001756-10EA
VT001753-10EAEA 124 EUR
VT001753-10EA VT001754-10EA VT001756-10EA
Klebsiella aerogenes (formerly Enterobacter aerogenes) WDCM 00175 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Padrões Biological Standards
Vitroids™ discs contain Klebsiella aerogenes WDCM 00175 viable microorganisms in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.
  • Defined CFU range and low standard deviation
  • Fast, reliable, and easy to use
The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year, and are in a viable state with a shelf life of 1-3 years. Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data, and the number of passages (subcultures) from the original strain.
Vitroids™ are derived from a traceable culture obtained freeze-dried from CECT®.
Certificações: These certified reference materials (CRMs) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025.