Print…
VWR®, Side Access Rack for Cryoboxes, Height 50 mm
Fornecedor: VWR Collection
479-2265EA 97.7 EUR
479-2265 479-2266 479-2267 479-2268 479-2269 479-2270 479-2271
VWR®, Side Access Rack for Cryoboxes, Height 50 mm
Suportes/Racks Suportes de caixas criogénicas
Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
  • Different height and depth dimensions are available and can be used in all freezer brands
  • Racks for standard 2" cryoboxes (133×133×51 mm)
  • Made from corrosion resistant stainless steel with smoothed edges
  • Organise and find your samples quickly
These racks are made from corrosion resistant stainless steel with smoothed edges. Side access racks are a cost-effective freezer rack that will fit into any freezer.
Measure the inside height and depth in the compartment of your freezer before choosing the right racks. In 'Related information' tab you will find additional information to support you in the selection.
Informações de encomenda: Delivered without cryoboxes.