Print…
VWR®, Drawer Rack for Cryoboxes, Height 50 mm
Fornecedor: VWR Collection
479-2258EA 167 EUR
479-2258 479-2259 479-2260 479-2261 479-2262 479-2263 479-2264
VWR®, Drawer Rack for Cryoboxes, Height 50 mm
Suportes/Racks Suportes de caixas criogénicas
Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
  • Different height and depth dimensions are available and can be used in all freezer brands
  • Racks for standard 2" cryoboxes (133×133×51 mm)
  • Made from corrosion resistant stainless steel with smoothed edges
These racks ensure an ergonomic operating process as the drawers are placed in the rack. With a pull-out design it makes it easy to get samples without taking the whole rack out from the freezer. The tray has an easy access handle.
Measure the inside height and depth in the compartment of your freezer before choosing the right racks. In 'Related information' tab you will find additional information to support you in the selection.
Informações de encomenda: Delivered without cryoboxes.