Print…
VWR® Sliding Tray Racks for Cryoboxes, Height 50 mm
Fornecedor: VWR Collection
479-2251EA 163 EUR
479-2251 479-2252 479-2253 479-2254 479-2255 479-2256 479-2257 479-2437
VWR® Sliding Tray Racks for Cryoboxes, Height 50 mm
Suportes/Racks Suportes de caixas criogénicas
Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
  • Different height and depth dimensions are available and can be used in all freezer brands
  • Racks for standard 2" cryoboxes (133×133×51 mm)
  • Made from corrosion resistant Stainless steel with smoothed edges
These racks ensure an ergonomic operating process as the trays are placed in the rack. With a pull-out design it makes it easy to get samples without taking the whole rack out from the freezer. The tray has an easy access handle.
Measure the inside height and depth in the compartment of your freezer before choosing the right racks. In 'Related information' tab you will find additional information to support you in the selection.
Informações de encomenda: Delivered without cryoboxes.