Print…
Removable deep-freeze labels for microplates
Fornecedor: GA
AMA-11C1-2WH AMA-11C3-4WH AMA-223C1-2BL AMA-223C1-2GA AMA-223C1-2WH AMA-223C1-2YE AMA-223C3-4BL AMA-223C3-4GA AMA-223C3-4WH AMA-223C3-4YE
GAINAMA-11C1-2WHEA 90.6 EUR
GAINAMA-11C1-2WH GAINAMA-11C3-4WH GAINAMA-223C1-2BL GAINAMA-223C1-2GA GAINAMA-223C1-2WH GAINAMA-223C1-2YE GAINAMA-223C3-4BL GAINAMA-223C3-4GA GAINAMA-223C3-4WH GAINAMA-223C3-4YE
Removable deep-freeze labels for microplates
Rótulos
These micro-well plate labels are particularly useful for microbiology, genetics, and drug discovery laboratories.
  • Perfectly suited for the identification of microtiter plates
  • Able to withstand a wide temperature range: –80 to 72 °C (–112 to 161,6 °F)
  • Glove-friendly adhesive ensures labels can be removed without leaving any glue residue behind
Waterproof labels made of a durable thermoplastic film are ideal for use on cell culture plates and flasks, micro-well plates, as well as PCR plates and Petri dishes. These labels retain their shape when removed from the liner, making them easy to manipulate. This design facilitates the application of the label to any multiwell plate or Petri dish. Their glove-friendly, removable adhesive makes the label easy to handle and reposition, while still providing a strong adhesion in incubators or freezers. They are perfect for everyday uses and long-term freezer storage.