Print…
Ice crusher, self contained, Ice Queen
Fornecedor: ITVA
ICEQ85CAIR ICEQ135AIR
474-0089EA 6120 EUR
474-0089 474-0087
Ice crusher, self contained, Ice Queen
Trituradores de gelo
ICE QUEEN range of crushed-ice produce granular ice with a vertical cylindrical evaporator and heavy duty auger mechanism. This range has been specially desiged for areas with high content in lime and difficult conditions.
  • Simple to use
  • Its electronic-free design makes it ideal for water areas abd toughest working conditions
  • Body easy to remove
  • Its oversized condenser is optimum for hot climates
  • The water content of the ice can be adjusted to suit every customer's requirements, it means that the machine can be adjusted to produce dumper or drier ice
Fornecimento: The ICE QUEEN models are supplied complete with water filter, scoop and ice distributor