Print…
Multiple distributors for bottles with GLS 80 thread, BOLA
Fornecedor: Bohlender
D754-16 D754-24
201-0293EA 126 EUR
201-0293 BOHLD754-24
Multiple distributors for bottles with GLS 80 thread, BOLA
Tampas Tampas para Frascos
PPS screw cap, blue, distributor body made from PTFE.
  • Very good chemical resistance
  • Temperature resistant from –200 to +250 °C
  • Suitable for vacuum applications
For the removal or filling of aggressive or pure liquids. For the connection of hoses, tubes and probes into vessels. Hoses with an external diameter of maximum 12,7 mm can be inserted through the stopcocks.