Print…
Gamma irradiated cleanroom markers, Sharpie®
Fornecedor: KEYSTONE CLEANROOM PRODUCTS
S01UFMARKBLK S01UFMARKBLUE S01FMARKBLK S01FMARKBLUE S01RETMARKBLK S01RETMARKBLUE S02UFMARKBLK S02UFMARKBLUE S02FMARKBLK S02FMARKBLUE S05UFMARKBLK S05UFMARKBLUE S05FMARKBLK S05FMARKBLUE S10UFMARKBLK S10UFMARKBLUE S10FMARKBLK S10FMARKBLUE
129-3113EA 3220 EUR
129-3113 129-3114 129-3105 129-3106 129-3109 129-3110 129-3119 129-3120 129-3115 129-3116 129-2942 129-2943 129-3121 129-3057 129-2948 129-2949 129-2944 129-2945
Gamma irradiated cleanroom markers, Sharpie®
Canetas Cleanroom Pens
These pens and markers eliminate the risks associated with surface sanitisation procedures.
  • Lot controlled
  • Ready to use
  • Packed in easy to open Tyvek® peel pouches
  • Minimises contamination risks
  • Simplifies conformance to SOP's
  • Eliminates the need for wiping down contaminated writing instruments
CRP offers gamma irradiated pens and markers for the convenience of pharmaceutical cleanroom operations. These are a 'Ready to Use Option' when sterilisation on-site is not possible; or surface sterilisation is insufficient. The clean Tyvek® peel pouch packaging allows for effective surface decontamination and easy introduction into the cleanroom through the material handling airlock.
Certificações: Certificates of Irradiation and Conformance with each shipment.