Print…

Fluorocyclopentane

Fornecedor: Apollo Scientific
Atenção

PC3147-25G PC3147-5G
APOSPC3147-25GEA 693 EUR
APOSPC3147-25G APOSPC3147-5G
Fluorocyclopentane
Fluorocyclopentane
Fórmula: C₅H₉F
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD08059532
Número CAS: 1481-36-3
UN: 1993
ADR: 3 II

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR