Print…

Dicyclopropylmethanamine 98%

Fornecedor: Apollo Scientific
Atenção

OR940675-1G OR940675-25G OR940675-5G
APOSOR940675-1GEA 68.4 EUR
APOSOR940675-1G APOSOR940675-25G APOSOR940675-5G
Dicyclopropylmethanamine 98%
Dicyclopropylmethanamine
Fórmula: C₇H₁₃N
Peso Molecular: 111.19 g/mol
Temperatura de Armazenamento: Frigorífico
Número MDL : MFCD06804453
Número CAS: 13375-29-6

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR