Ethyl-N-hydroxyacetimidate 96%
Fornecedor: Apollo Scientific
Sinônimos:
Ethyl acetohydroximate
OR931234-100G
OR931234-25G
APOSOR931234-100GEA
171
EUR
APOSOR931234-100G
APOSOR931234-25G
Ethyl-N-hydroxyacetimidate 96%
Ethyl-N-hydroxyacetimidate
Fórmula:
C₄H₉NO₂ Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD00002114 Número CAS: 10576-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...