Print…

N-Cyano-N',S-dimethylisothiourea 95%

Fornecedor: Apollo Scientific

Sinônimos: Methyl N'-cyano-N-methylcarbamimidothioate

OR920511-25G OR920511-5G
APOSOR920511-25GEA 382 EUR
APOSOR920511-25G APOSOR920511-5G
N-Cyano-N',S-dimethylisothiourea 95%
N-Cyano-N',S-dimethylisothiourea
Fórmula: C₄H₇N₃S
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00078154
Número CAS: 5848-24-8

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR