Print…

Allyl benzoate 95%

Fornecedor: Apollo Scientific

Sinônimos: Benzoic acid Allyl Ester, Benzoic Acid Allyl Ester, Allyl Benzoate

OR919704-100G OR919704-25G
APOSOR919704-100GEA 200 EUR
APOSOR919704-100G APOSOR919704-25G
Allyl benzoate 95%
Allyl benzoate
Fórmula: C₁₀H₁₀O₂
Peso Molecular: 162.19 g/mol
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00026104
Número CAS: 583-04-0

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR