Print…

2-Hydroxyterephthalic acid dimethyl ester

Fornecedor: Apollo Scientific
Atenção

OR6979-1G OR6979-5G
APOSOR6979-1GEA 0 EUR
APOSOR6979-1G APOSOR6979-5G
2-Hydroxyterephthalic acid dimethyl ester
2-Hydroxyterephthalic acid dimethyl ester
Fórmula: C₁₀H₁₀O₅
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD08445666
Número CAS: 6342-72-9

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR