(+)-Isomenthol 95%
Fornecedor: Apollo Scientific
|
Atenção
|
OR480700-5G
OR480700-250MG
OR480700-1G
APOSOR480700-5GEA
0
EUR
APOSOR480700-5G
APOSOR480700-250MG
APOSOR480700-1G
(+)-Isomenthol 95%
(+)-Isomenthol
Fórmula:
C₁₀H₂₀O Temperatura de Armazenamento: Frigorífico |
Número CAS:
23283-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...