Isopropoxyacetic acid 98%
Fornecedor: Apollo Scientific
|
Atenção
|
OR46564-1G
OR46564-250MG
OR46564-5G
APOSOR46564-1GEA
0
EUR
APOSOR46564-1G
APOSOR46564-250MG
APOSOR46564-5G
Isopropoxyacetic acid 98%
Isopropoxyacetic acid
Fórmula:
C₅H₁₀O₃ Temperatura de Armazenamento: Ambiente |
Número MDL :
MFCD09055367 Número CAS: 33445-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...