Print…
O-Methylhydroxylamine hydrochloride 98%
Fornecedor: Apollo Scientific
Perigo
Sinônimos: Methylhydroxylamine hydrochloride, O-Methylhydroxylammonium chloride, Methoxyamine hydrochloride, Methoxylamine hydrochloride
OR18305-100G OR18305-25G OR18305-500G
APOSOR18305-100GEA 43.5 EUR
APOSOR18305-100G APOSOR18305-25G APOSOR18305-500G
O-Methylhydroxylamine hydrochloride 98%
O-Methylhydroxylamine hydrochloride
Fórmula: CH₆ClNO
Peso Molecular: 83.515 g/mol
Ponto de Fusão: 147…154 °C (decomposição)
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00012951
Número CAS: 593-56-6
EINECS: 209-798-7
UN: 3261
ADR: 8 II