2-(4-Methylphenyl)-2-phenylacetic acid
Fornecedor: Apollo Scientific
|
Atenção
|
OR12757-250MG
OR12757-1G
APOSOR12757-250MGEA
531
EUR
APOSOR12757-250MG
APOSOR12757-1G
2-(4-Methylphenyl)-2-phenylacetic acid
2-(4-Methylphenyl)-2-phenylacetic acid
Fórmula:
C₁₅H₁₄O₂ Temperatura de Armazenamento: Ambiente |
Número CAS:
1882-56-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...